Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroxyacetone, Technical Grade

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16034-1000 16034-5000
TS16034-1000EA 44 USD InStock
TS16034-1000 TS16034-5000
Hydroxyacetone, Technical Grade
Hydroxyacetone


Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 145
Melting Pt: -17
Density: 1.08 g/cm³
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004669
CAS Number: 116-09-6
UN: 1224
ADR: 3 III
Merck Index: 13,00066
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty