Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitrobenzenesulfenyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16133-0100 16133-0500
TS16133-0100EA 66 USD InStock
TS16133-0100 TS16133-0500
2-Nitrobenzenesulfenyl chloride 98%
2-Nitrobenzenesulphenyl chloride


Formula: C₆H₄ClNO₂S
MW: 189.62 g/mol
Melting Pt: 70…75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007128
CAS Number: 7669-54-7
EINECS: 231-644-2
UN: 1759
ADR: 8 II
Order Now
Specification Test Results
Appearance Yellow to golden Crystalline powder or needles
Infrared spectrum Conforms
Melting point 70 to 75 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty