Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-p-anisaldehyde 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16257-0050 16257-0250 16257-1000
TS16257-0050EA 26 USD InStock
TS16257-0050 TS16257-0250 TS16257-1000
3-Hydroxy-p-anisaldehyde 98%
3-Hydroxy-p-anisaldehyde


Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 308
Melting Pt: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9
Order Now
Specification Test Results
Appearance Off-white to grey to light brown Powder
Infrared spectrum Conforms
Melting point 112 to 116 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty