Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Bromobutyric acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16451-0010 16451-0050 16451-2500
TS16451-0010EA 392 USD InStock
TS16451-0010 TS16451-0050 TS16451-2500
(±)-2-Bromobutyric acid 99%
(±)-2-Bromobutyric acid


Formula: H₃CCH₂BrCHCOOH
MW: 167 g/mol
Boiling Pt: 214
Melting Pt: -4
Density: 1.56 g/cm³
Flash Pt: >112 °C
MDL Number: MFCD00004216
CAS Number: 80-58-0
EINECS: 201-294-5
UN: 2922
ADR: 8 II
Merck Index: 13,01397
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4715 to 1.4755 (20 °C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty