Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Oxoglutaric acid 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17315-1000 17315-5000
TS17315-1000EA 84 USD InStock
TS17315-1000 TS17315-5000
3-Oxoglutaric acid 96%
3-Oxoglutaric acid


Formula: C₅H₆O₅
MW: 146.1 g/mol
Boiling Pt: 323
Melting Pt: 138 (dec.)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002711
CAS Number: 542-05-2
EINECS: 208-797-9
Merck Index: 13,00069
Order Now
Specification Test Results
Appearance White to beige Powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
Titration with NaOH ≥95 %
Sulfuric acid ≤2 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty