Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dihydroxybenzaldehyde 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17364-0250 17364-1000
TS17364-0250EA 68 USD InStock
TS17364-0250 TS17364-1000
2,4-Dihydroxybenzaldehyde 98%
2,4-Dihydroxybenzaldehyde


Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 351
Melting Pt: 133…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00011686
CAS Number: 95-01-2
EINECS: 202-383-1
Merck Index: 12,08324
Order Now
Specification Test Results
Appearance Light beige to pink-orange Powder
Infrared spectrum Conforms
Melting point 134 to 140 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty