Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxybenzaldehyde 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17425-0250 17425-1000
TS17425-0050EA 23.69 USD InStock
TS17425-0050 TS17425-0250 TS17425-1000
4-Ethoxybenzaldehyde 99%
4-Ethoxybenzaldehyde


Formula: C₉H₁₀O₂
Boiling Pt: 255 °C (760 mmHg)
Melting Pt: 13…14 °C
Density: 1.08 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00003388
CAS Number: 10031-82-0
EINECS: 233-093-3
Order Now
Specification Test Results
Appearance Clear yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5574 to 1.5594 (20 °C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty