Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

exo-2-Aminonorbornane 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17695-0010 17695-0050
TS17695-0010EA 75 USD InStock
TS17695-0010 TS17695-0050
exo-2-Aminonorbornane 99%
exo-2-Aminonorbornane


Formula: C₇H₁₃N
Boiling Pt: 49 °C (13mbar)
Density: 0.93 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00078132
CAS Number: 7242-92-4
UN: 2733
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty