Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methylisatin 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17935-0010 17935-0100
TS17935-0010EA 39 USD InStock
TS17935-0010 TS17935-0100
1-Methylisatin 98%
1-Methylisatin


Formula: C₉H₇NO₂
MW: 161.16
Melting Pt: 139
Density: 1.4
Storage Temperature: Ambient
MDL Number: MFCD00005812
CAS Number: 2058-74-4
EINECS: 218-164-9
Order Now
Specification Test Results
Appearance Orange to brown Crystalline powder
Infrared spectrum Conforms
Melting point 129 to 133 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty