Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Naphthalenemethanol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

18011-0020 18011-0100
TS18011-0020EA 29 USD InStock
TS18011-0020 TS18011-0100
1-Naphthalenemethanol 98%
1-Naphthalenemethanol


Formula: C₁₁H₁₀O
Boiling Pt: 301 °C (715 mmHg)
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00004044
CAS Number: 4780-79-4
EINECS: 225-324-1
Order Now
Specification Test Results
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 59 to 62 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty