Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl adipate 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

18131-1000 18131-5000
TS18131-1000EA 34 USD InStock
TS18131-1000 TS18131-5000
Dimethyl adipate 99+%
Dimethyl adipate


Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 229
Melting Pt: 10.3
Density: 1.06 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00008469
CAS Number: 627-93-0
EINECS: 211-020-6
Merck Index: 13,00163
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4290 (20 °C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.0 %
Color scale ≤20 APHA
Acidity ≤0.01 % (as adipic acid)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty