Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Caproic anhydride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

18675-0500 18675-2500
TS18675-0500EA 23 USD InStock
TS18675-0500 TS18675-2500
Caproic anhydride 97%
Caproic anhydride


Formula: C₁₂H₂₂O₃
MW: 214.3 g/mol
Boiling Pt: 246
Melting Pt: -40
Density: 0.926 g/cm³
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00009509
CAS Number: 2051-49-2
EINECS: 218-121-4
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4300 (20 °C, 589 nm)
Titration by coupling ≥96.0 %
HPLC ≥97.5 %
Sodium (Na) passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty