Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isopropylbenzyl alcohol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

18780-0250 18780-1000 18780-5000
TS18780-0250EA 35 USD InStock
TS18780-0250 TS18780-1000 TS18780-5000
4-Isopropylbenzyl alcohol 97%
4-Isopropylbenzyl alcohol


Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 249
Melting Pt: 28
Density: 0.98 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004663
CAS Number: 536-60-7
EINECS: 208-640-4
Merck Index: 13,02646
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5180 to 1.5210 (20 °C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty