Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1H-Imidazol-4-yl)methanol hydrochloride 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20487-0010 20487-0050
TS20487-0010EA 60 USD InStock
TS20487-0010 TS20487-0050
(1H-Imidazol-4-yl)methanol hydrochloride 99%
(1H-Imidazol-4-yl)methanol hydrochloride


Formula: C₄H₇ClN₂O
MW: 134.57 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00012697
CAS Number: 32673-41-9
EINECS: 251-150-0
Order Now
Specification Test Results
Appearance Light yellow to beige to light brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty