Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methylisatin 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20686-0010 20686-0050
TS20686-0010EA 36 USD InStock
TS20686-0010 TS20686-0050
5-Methylisatin 95%
5-Methylisatin


Formula: C₉H₇NO₂
Melting Pt: 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005721
CAS Number: 608-05-9
Order Now
Specification Test Results
Appearance Orange-red to red-purple to brown Crystalline powder
Infrared spectrum Conforms
Melting point 186 to 190 °C
HPLC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty