Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pentynoic acid 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

21373-0010 21373-0050
TS21373-0010EA 64 USD InStock
TS21373-0010 TS21373-0050
4-Pentynoic acid 95%
4-Pentynoic acid


Formula: C₅H₆O₂
Boiling Pt: 110 °C (30 mmHg)
Melting Pt: 54…58 °C
Flash Pt: 75 °C
MDL Number: MFCD00004407
CAS Number: 6089-09-4
EINECS: 228-028-0
UN: 2923
ADR: 8,II
Order Now
Specification Test Results
Appearance White to yellow to beige to light brown Crystalline powder or flakes and/or chunks
Infrared spectrum Conforms
Melting point 54 to 60 °C
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty