Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

21807-0050 21807-0250
TS21807-0050EA 108 USD InStock
TS21807-0050 TS21807-0250
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol


Formula: C₆H₁₂O₃
Boiling Pt: 82…83 °C (14 mmHg)
Density: 1.07 g/cm³
Flash Pt: 80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063239
CAS Number: 22323-82-6
EINECS: 244-910-8
Merck Index: 12,05232
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20 °C, 589 nm)
GC ≥97.5 %
Specific optical rotation +10.5° to +11.5° (20°C, 589 nm) (c=5, CH3OH)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty