Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylbutanone 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

21877-0010 21877-0051 21877-2500
TS21877-0010EA 186 USD InStock
TS21877-0010 TS21877-0051 TS21877-2500
3-Methylbutanone 98%
3-Methylbutanone


Formula: C₅H₁₀O
Boiling Pt: 94 °C (1013 hPa)
Density: 0.8 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00008919
CAS Number: 563-80-4
EINECS: 209-264-3
UN: 2397
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty