Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenylpropanal 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

24051-0050 24051-1000 24051-5000
TS24051-0050EA 49 USD InStock
TS24051-0050 TS24051-1000 TS24051-5000
3-Phenylpropanal 95%
3-Phenylpropanal


Formula: C₉H₁₀O
Boiling Pt: 224…226 °C (760 mmHg)
Melting Pt: –42 °C
Density: 1.01 g/cm³
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00007021
CAS Number: 104-53-0
EINECS: 203-211-8
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5300 (20 °C, 589 nm)
GC ≥94.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty