Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isobutyric anhydride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

25778-0020 25778-5000
TS25778-0020EA 294 USD InStock
TS25778-0020 TS25778-5000
Isobutyric anhydride 97%
Isobutyric anhydride


Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 183…187 °C (1013 hPa)
Melting Pt: -69
Density: 0.954 g/cm³
Flash Pt: 66.4 °C
MDL Number: MFCD00008913
CAS Number: 97-72-3
EINECS: 202-603-6
UN: 2922
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20 °C, 589 nm)
GC ≤1.0 % (Isobutyric acid)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty