Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl stearate mixture of homologs

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

30422-0010 30422-0025
TS30422-0010EA 80 USD InStock
TS30422-0010 TS30422-0025
Methyl stearate mixture of homologs
Methyl stearate


Formula: CH₃(CH₂)₁₆CO₂CH₃
MW: 298.51 g/mol
Boiling Pt: 355
Melting Pt: 37…39 °C
Density: 0.836 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009005
CAS Number: 112-61-8
Merck Index: 12,08959
Order Now
Specification Test Results
Appearance White to cream Waxy solid
Infrared spectrum Conforms
Acid value ≤1 mg KOH/g
Iodine value ≤1 g I2/100 g
Saponification value 186 to 208 mg KOH/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty