Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Butynoic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30806-0010 30806-0050
TS30806-0010EA 49 USD InStock
TS30806-0010 TS30806-0050
2-Butynoic acid 98%
2-Butynoic acid


Formula: C₄H₄O₂
MW: 84.07
Boiling Pt: 203
Melting Pt: 78
Density: 0.9641
Storage Temperature: Ambient
MDL Number: MFCD00004363
CAS Number: 590-93-2
EINECS: 209-695-7
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 74 to 78 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty