Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-2-Methoxypropionic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

33172-0025 33172-5000
TS33172-0025EA 533 USD InStock
TS33172-0025 TS33172-5000
(S)-(-)-2-Methoxypropionic acid 98%
(S)-(-)-2-Methoxypropionic acid


Formula: C₄H₈O₃
MW: 104.105 g/mol
Boiling Pt: 99 °C (20 mmHg)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD01632589
CAS Number: 23953-00-6
UN: 3265
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4132 to 1.4152 (20 °C, 589 nm)
GC ≥97.5 %
Specific optical rotation -70° to -80° (20°C, 589 nm) (neat)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty