Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetylbenzenesulphonyl chloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

34385-0050 34385-0500
TS34385-0050EA 72 USD InStock
TS34385-0050 TS34385-0500
4-Acetylbenzenesulphonyl chloride 97%
4-Acetylbenzenesulphonyl chloride


Formula: C₈H₇ClO₃S
MW: 218.66
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00800269
CAS Number: 1788-10-9
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Appearance White to orange to brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 83 to 87 °C
Additional info product may contain varying amounts of inorganic chloride
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty