Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-1-Phenylethanol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35188-1000 35188-5000
TS35188-1000EA 35 USD InStock
TS35188-1000 TS35188-5000
(±)-1-Phenylethanol 97%
(±)-1-Phenylethanol


Formula: C₆H₅CH(OH)CH₃
MW: 122.17 g/mol
Boiling Pt: 401
Melting Pt: 20
Density: 1.01 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00004508
CAS Number: 98-85-1
EINECS: 202-707-1
UN: 2937
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5290 (20 °C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty