Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetylphenylboronic acid 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35882-0010 35882-0050 35882-0250
TS35882-0010EA 56 USD InStock
TS35882-0010 TS35882-0050 TS35882-0250
2-Acetylphenylboronic acid 96%
2-Acetylphenylboronic acid


Formula: C₈H₉BO₃
Melting Pt: 170 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD01321263
CAS Number: 308103-40-4
Order Now
Specification Test Results
Appearance White to yellow Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty