Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Bromo-4-cyanoacetophenone 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

36536-0050 36536-0250
TS36536-0050EA 41 USD InStock
TS36536-0050 TS36536-0250
ω-Bromo-4-cyanoacetophenone 97%
ω-Bromo-4-cyanoacetophenone


Formula: C₉H₆BrNO
MW: 224.06
Boiling Pt: 314
Melting Pt: 85…88 °C
MDL Number: MFCD00052931
CAS Number: 20099-89-2
UN: 2928
ADR: 6.1 II
Order Now
Specification Test Results
Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 96 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty