Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoroisatin 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36770-0050 36770-0250
TS36770-0050EA 67 USD InStock
TS36770-0050 TS36770-0250
5-Fluoroisatin 98%
5-Fluoroisatin


Formula: C₈H₄FNO₂
MW: 165.12 g/mol
Melting Pt: 224…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00022795
CAS Number: 443-69-6
Order Now
Specification Test Results
Appearance Red to brown Powder
Infrared spectrum Conforms
Melting point 224 to 230 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty