Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetylphenylboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36940-0010 36940-0050 36940-0250
TS36940-0010EA 27 USD InStock
TS36940-0010 TS36940-0050 TS36940-0250
4-Acetylphenylboronic acid 97%
4-Acetylphenylboronic acid


Formula: C₈H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD01074667
CAS Number: 149104-90-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty