Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methylimidazole-2-carbaldehyde 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36985-0010 36985-0050
TS36985-0010EA 51 USD InStock
TS36985-0010 TS36985-0050
1-Methylimidazole-2-carbaldehyde 97%
1-Methylimidazole-2-carbaldehyde


Formula: C₅H₆N₂O
MW: 110.12 g/mol
Boiling Pt: 70…74 °C (1 mmHg)
Melting Pt: 35…39 °C
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD01321308
CAS Number: 13750-81-7
Order Now
Specification Test Results
Appearance White to yellow to orange Crystals or needles or crystalline mass and/or chunks
Infrared spectrum Conforms
Melting point 35 to 39 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty