Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Quinolinecarbaldehyde 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

37017-0050 37017-0250
TS37017-0050EA 74 USD InStock
TS37017-0050 TS37017-0250
2-Quinolinecarbaldehyde 97%
2-Quinolinecarbaldehyde


Formula: C₁₀H₇NO
MW: 157.17
Melting Pt: 66…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075032
CAS Number: 5470-96-2
EINECS: 226-804-3
Order Now
Specification Test Results
Appearance Yellow to orange to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 66 to 71 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty