Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl benzoylacetate 90%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

37085-0050 37085-0500 37085-1000 37085-5000
TS37085-0050EA 29 USD InStock
TS37085-0050 TS37085-0500 TS37085-1000 TS37085-5000
Ethyl benzoylacetate 90%
Ethyl benzoylacetate


Formula: C₁₁H₁₂O₃
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.106 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00009196
CAS Number: 94-02-0
EINECS: 202-295-3
Merck Index: 13,03803
Order Now
Specification Test Results
Appearance Clear colorless to yellow to red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5240 to 1.5330 (20 °C, 589 nm)
HPLC ≥88 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty