Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-Epoxysuccinic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

37264-0050 37264-0250
TS37264-0050EA 120 USD InStock
TS37264-0050 TS37264-0250
cis-Epoxysuccinic acid 97%
cis-Epoxysuccinic acid


Formula: C₄H₄O₅
MW: 132.07 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00191645
CAS Number: 16533-72-5
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH 96.0 to 104.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty