Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl propargyl ether 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

38091-0010 38091-0050
TS38091-0010EA 32 USD InStock
TS38091-0010 TS38091-0050
tert-Butyl propargyl ether 97%
tert-Butyl propargyl ether


Formula: C₇H₁₂O
MW: 112.172 g/mol
Boiling Pt: 119 °C (760 mmHg)
Density: 0.835 g/cm³
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD07368651
CAS Number: 16314-18-4
UN: 3271
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20 °C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty