Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Mercapto-1-propanol 97+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

38857-0010 38857-0050
TS38857-0010EA 75 USD InStock
TS38857-0010 TS38857-0050
3-Mercapto-1-propanol 97+%
3-Mercapto-1-propanol


Formula: C₃H₈OS
Boiling Pt: 106 °C (50 mmHg)
Density: 1.062 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00192262
CAS Number: 19721-22-3
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4970 (20 °C, 589 nm)
GC ≥97.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty