Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-5-carbaldehyd 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39163-0010 39163-0050 39163-0250
TS39163-0010EA 43 USD InStock
TS39163-0010 TS39163-0050 TS39163-0250
Indole-5-carbaldehyd 98+%
Indole-5-carbaldehyd


Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD02093664
CAS Number: 1196-69-6
Order Now
Specification Test Results
Appearance Orange to tan Powder
Infrared spectrum Conforms
Melting point 98 to 103 °C
HPLC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty