Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylsalicylaldehyde 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39854-0010 39854-0050
TS39854-0010EA 38 USD InStock
TS39854-0010 TS39854-0050
3-Methylsalicylaldehyde 98%
3-Methylsalicylaldehyde


Formula: C₈H₈O₂
Boiling Pt: 203…205 °C (730 mmHg)
Density: 1.134 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00040798
CAS Number: 824-42-0
Order Now
Specification Test Results
Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5620 to 1.5650 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty