Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di-tert-butyl malonate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

40662-0100 40662-0500
TS40662-0100EA 71 USD InStock
TS40662-0100 TS40662-0500
Di-tert-butyl malonate 98%
Di-tert-butyl malonate


Formula: C₁₁H₂₀O₄
Boiling Pt: 110…111 °C (22 mmHg)
Melting Pt: –7 °C
Density: 0.966 g/cm³
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008810
CAS Number: 541-16-2
EINECS: 208-769-6
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4175 to 1.4195 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty