Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glyoxylic acid 50% in water

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

41106-0025 41106-0050 41106-5000
TS41106-0025EA 259 USD InStock
TS41106-0025 TS41106-0050 TS41106-5000
Glyoxylic acid 50% in water
Glyoxylic acid


Formula: C₂H₂O₃
Melting Pt: –93 °C
Storage Temperature: Ambient
MDL Number: MFCD00006958
CAS Number: 298-12-4
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Solution
Refractive index 1.4130 to 1.4200 (20°C, 589 nm)
Titration with NaOH ≥50 %
Enantiomeric excess ≥97.5 % (HPLC)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty