Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Hydroxyacetophenone 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

41191-0050 41191-1000
TS41191-0050EA 20 USD InStock
TS41191-0050 TS41191-1000
3'-Hydroxyacetophenone 99+%
3'-Hydroxyacetophenone


Formula: C₈H₈O₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00002298
CAS Number: 121-71-1
EINECS: 204-494-0
Order Now
Specification Test Results
Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 97 °C
GC ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty