Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4,4-dimethyl-3-oxovalerate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

41419-0010 41419-0100
TS41419-0010EA 24 USD InStock
TS41419-0010 TS41419-0100
Methyl-4,4-dimethyl-3-oxovalerate 98%
Methyl-4,4-dimethyl-3-oxovalerate


Formula: C₈H₁₄O₃
Boiling Pt: 67…70 °C (13 mmHg)
Density: 0.98 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00008847
CAS Number: 55107-14-7
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %
HPLC ≥97.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty