Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-Phenyl-1,3-propanediol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

41775-0010 41775-0050
TS41775-0010EA 133 USD InStock
TS41775-0010 TS41775-0050
(R)-1-Phenyl-1,3-propanediol 98%
(R)-1-Phenyl-1,3-propanediol


Formula: C₉H₁₂O₂
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00145213
CAS Number: 103548-16-9
Order Now
Specification Test Results
Appearance White Crystalline solid
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥97.5 %
Specific optical rotation +53° to +56° (22 °C, 589 nm) (c=1, 1,2-dichlorethane)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty