Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S)-1-Phenylpropane-1,3-diol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

41776-0010 41776-0050 41776-2500
TS41776-0010EA 239 USD InStock
TS41776-0010 TS41776-0050 TS41776-2500
(1S)-1-Phenylpropane-1,3-diol 98%
(1S)-1-Phenylpropane-1,3-diol


Formula: C₉H₁₂O₂
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00221510
CAS Number: 96854-34-1
Order Now
Specification Test Results
Appearance White Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥97.5 %
Specific optical rotation -53° to -56° (22 °C, 589 nm) (c=1, 1,2-dichlorethane)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty