Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,4-Trimethyl-1,3-pentanediol diisobutyrate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

42200-0010 42200-0050
TS42200-0010EA 56 USD InStock
TS42200-0010 TS42200-0050
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate 98%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate


Formula: C₁₆H₃₀O₄
MW: 286.41 g/mol
Boiling Pt: 280 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.94 g/cm³
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00059267
CAS Number: 6846-50-0
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty