Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-oxo-3-(pyridin-4-yl)propanoate 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

42548-0025 42548-0100
TS42548-0025EA 83 USD InStock
TS42548-0025 TS42548-0100
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate 95%
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate


Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
Melting Pt: 53…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00094021
CAS Number: 26377-17-3
Order Now
Specification Test Results
Appearance Off-white to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 59 °C
HPLC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty