Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl chlorophosphate 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

42614-0050 42614-0250
TS42614-0050EA 63 USD InStock
TS42614-0050 TS42614-0250
Dimethyl chlorophosphate 96%
Dimethyl chlorophosphate


Formula: C₂H₆ClO₃P
MW: 144.49 g/mol
Boiling Pt: 80 °C (25 mmHg)
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00117904
CAS Number: 813-77-4
UN: 2927
ADR: 6.1 I
Order Now
Specification Test Results
Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4140 (20 °C, 589 nm)
Titration Argentometric ≥95.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty