Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-2-carbaldehyd 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42751-0010 42751-0050
TS42751-0010EA 124 USD InStock
TS42751-0010 TS42751-0050
Indole-2-carbaldehyd 97%
Indole-2-carbaldehyd


Formula: C₉H₇NO
MW: 145.16
Melting Pt: 138…142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001425
CAS Number: 19005-93-7
Order Now
Specification Test Results
Appearance White to yellow to tan Crystalline powder
Infrared spectrum Conforms
Melting point 133 to 143 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty