Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hexanoate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43754-0100 43754-0250
TS43754-0100EA 81 USD InStock
TS43754-0100 TS43754-0250
Sodium hexanoate 99%
Sodium hexanoate


Formula: C₆H₁₁NaO₂
Melting Pt: 231.2…233.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00059056
CAS Number: 10051-44-2
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
Titration with HClO4 ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty