Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Pyridinedicarboxaldehyde 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43775-0010 43775-2500
TS43775-0010EA 296 USD InStock
TS43775-0010 TS43775-2500
2,6-Pyridinedicarboxaldehyde 97%
2,6-Pyridinedicarboxaldehyde


Formula: C₇H₅NO₂
Melting Pt: 123.3…124.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00010103
CAS Number: 5431-44-7
Order Now
Specification Test Results
Appearance White to tan Powder
Infrared spectrum Conforms
Melting point 122 to 126 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty