Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl benzenesulfinate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43861-0050 43861-0250
TS43861-0050EA 106 USD InStock
TS43861-0050 TS43861-0250
Methyl benzenesulfinate 98%
Methyl benzenesulphinate


Formula: C₇H₈O₂S
Boiling Pt: 110…112 °C (16Torr)
Density: 1.194 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00674051
CAS Number: 670-98-4
Order Now
Specification Test Results
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty